All-Weather Rain Gauge
Overflow chamber catches excess for later measurement.
Supplier:  Customer Channels Group (Ccg) 88994
Catalog No. S63623
Explore available promotions
Includes:
  • Mounting bracket
  • One-year rainfall record
Description
Description
  • Graduated for metric measurement
  • Rain funnels into a removable inner tube
Specifications
Specifications
Mounting bracket, one year rainfall record
Rain Gauge
Globe Program