missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allantoin, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 102841000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allantoin | |
97-59-6 | |
100.0 | |
White | |
98% | |
C4H6N4O3 | |
100 g | |
15, 250 | |
Solubility in water: 1g/190mL. Other solubilities: readily soluble in alkalies,1g/500mL alcohol,practically insoluble in ether | |
C1(C(=O)NC(=O)N1)NC(=O)N | |
158.12 | |
CHEBI:15676 | |
98% |
98% | |
97.5 | |
230.0°C | |
Authentic | |
Plastic bottle | |
MFCD00005260 | |
25, V,15, 338 | |
allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream | |
POJWUDADGALRAB-UHFFFAOYSA-N | |
(2,5-dioxoimidazolidin-4-yl)urea | |
204 | |
158.12 | |
Powder |
Chemical Identifiers
97-59-6 | |
158.12 | |
POJWUDADGALRAB-UHFFFAOYSA-N | |
204 | |
(2,5-dioxoimidazolidin-4-yl)urea |
C4H6N4O3 | |
MFCD00005260 | |
allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream | |
CHEBI:15676 | |
C1(C(=O)NC(=O)N1)NC(=O)N |
Safety and Handling
EINECSNumber : 202-592-8
RTECSNumber : YT1600000
TSCA : TSCA
RUO – Research Use Only