missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allegro™ 16 in. Ducting
Supplier: Allegro™ 960025
Specifications
Yellow with black weather strip | |
Ducting | |
Vinyl and polyester with PVC coating |
16 in. | |
25 ft. |