Learn More
Allegro™ Banana Oil Fit-Test Kits
Isoamyl Acetate, also referred to as Banana Oil, is a qualitative fit test agent used to detect respirator leakage.
Supplier:  Allegro™ 0202
 Afficher plus de versions de ce produit
Numéro de catalogue. 19-088-504
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Banana oil (isoamyl acetate) is the substance originally used for testing respirator fit; its strong, recognizable odor quickly indicates the lack of proper fit
  • Available in two fit test kits and Fit Check Ampules that provide a fast and easy method of on-the-spot qualitative fit testing
Order Info
The Standard Kit comes in a handy carry bag and includes two 1 oz. (30mL) bottles of testing agent, a vinyl test enclosure, measuring pipette, and paper towels. The Deluxe Kit comes in a convenient storage case and contains everything necessary to perform a proper fit test, including an 8 1oz. (30mL) bottles of testing solution, a fit test tent, jars, paper towels, disposable bags, and other necessary components. Replacement banana oil solution is available in 1 oz. (30mL) bottles.
Spécifications
Spécifications
Deluxe Banana Oil Fit Test Kit
Fit test tent, jars, eight 1 oz. bottles of testing agent, paper towels, disposable bags and other necessary components, equipped in a convenient storage and carry case
1 Ea.
Respirator Fit Testing
0202
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS