Allegro™ Irritant Smoke Fit Test Kit
For qualitative fit testing of air purifying respirators.
Supplier: Allegro™ CAN2050
Description
- For qualitative fit testing of air purifying respirators
- Offers everything needed for meeting OSHA's protocol for fit testing with smoke
- Includes irritant smoke tubes, aspirator assembly, and record form in protective carry pouch
Specifications
Standard Smoke Test Kit | |
For qualitative fit testing of air purifying respirators | |
6 smoke tubes, aspirator bulb, tube tip breaker, record form, protective pouch | |
1 Ea. |
OSHA | |
Disposable or reusable respirators with p100 filter | |
2050 |