Learn More
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
$102.33 - $1414.78
Chemical Identifiers
CAS | 3237-50-1 |
---|---|
Molecular Formula | C4H4N2O5 |
Molecular Weight (g/mol) | 160.085 |
MDL Number | MFCD00149399 |
InChI Key | ZIIHZVKHFWOENY-UHFFFAOYSA-N |
Synonym | barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6 |
PubChem CID | 312231 |
IUPAC Name | 5,5-dihydroxy-1,3-diazinane-2,4,6-trione |
SMILES | C1(=O)C(C(=O)NC(=O)N1)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1532414
|
Thermo Scientific Chemicals
A1532414 |
25 g |
Each for $102.33
|
|
|||||
AAA1532418
|
Thermo Scientific Chemicals
A1532418 |
50 g |
Each for $168.86
|
|
|||||
AAA1532422
|
Thermo Scientific Chemicals
A1532422 |
100 g |
Each for $282.71
|
|
|||||
AAA1532430
|
Thermo Scientific Chemicals
A1532430 |
250 g |
Each for $551.01
|
|
|||||
AAA153240B
|
Thermo Scientific Chemicals
A153240B |
1000 g |
Each for $1,414.78
|
|
|||||
Description
Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.
Solubility
Soluble in water, ethanol, acetone, glacial acetic acid and methanol. Slightly soluble in chloroform, petroleum ether, toluene, ethyl acetate and acetic anhydride. Insoluble in ether.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
3237-50-1 | |
160.085 | |
ZIIHZVKHFWOENY-UHFFFAOYSA-N | |
312231 | |
C1(=O)C(C(=O)NC(=O)N1)(O)O |
C4H4N2O5 | |
MFCD00149399 | |
barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6 | |
5,5-dihydroxy-1,3-diazinane-2,4,6-trione |
Specifications
∼260°C (decomposition) | |
C4H4N2O5 | |
5309394 | |
barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6 | |
C1(=O)C(C(=O)NC(=O)N1)(O)O | |
160.085 | |
160.09 (142.07 Anhydrous) | |
25 g |
3237-50-1 | |
MFCD00149399 | |
14,282 | |
ZIIHZVKHFWOENY-UHFFFAOYSA-N | |
5,5-dihydroxy-1,3-diazinane-2,4,6-trione | |
312231 | |
98% | |
Alloxan monohydrate |
Safety and Handling
RTECSNumber : UW0492000
TSCA : TSCA
Recommended Storage : 2° to 8°C
RUO – Research Use Only