missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 2
Excellent for high-precision calibrations and reference standards
$2533.03 - $3709.47
Specifications
Product Type | Medium Metric Analytical Weight Set |
---|---|
Classification | ASTM Class 2 |
Packaging Quantity | Multiple |
Catalog Number | Mfr. No. | Weight (Metric) | Material | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Weight (Metric) | Material | Quantity | Price | Quantity | |||
02-217-1B
|
N/A | 1 to 100 g | Stainless Steel | 9/Set |
Each for $2,533.03
|
|
|||
02-215-116
|
Troemner™
80859247 |
1 mg to 200 g | Stainless Steel, Aluminum | 22/Set |
Each for $3,709.47
|
|
|||
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
- Low magnetic susceptibility, uniform consistency and high resistance to corrosion
- Used in extreme stable environments
- 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- Weight sets 1mg-500g are provided in a high quality polycarbonate set case
- Weight sets where the largest weight is 1kg and above are supplied in a felt line oak case
- Each polycarbonate case is placed in its own individual compartment
1Set includes two 200g weights. 2Set includes two 2kg weights.
Specifications
Medium Metric Analytical Weight Set | |
ASTM Class 2 | |
Multiple |