missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allura Red AC, TCI America™
$41.48 - $81.29
Chemical Identifiers
CAS | 25956-17-6 |
---|---|
Molecular Formula | C18H14N2Na2O8S2 |
Molecular Weight (g/mol) | 496.42 |
MDL Number | MFCD00059526 |
InChI Key | POXPUHKJGLHZEM-VIPPSAFOSA-L |
Synonym | FD and C Red No.40 |
PubChem CID | 44386827 |
IUPAC Name | disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate |
SMILES | [Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O |
Chemical Identifiers
25956-17-6 | |
496.42 | |
POXPUHKJGLHZEM-VIPPSAFOSA-L | |
44386827 | |
[Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O |
C18H14N2Na2O8S2 | |
MFCD00059526 | |
FD and C Red No.40 | |
disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate |
Specifications
25956-17-6 | |
MFCD00059526 | |
POXPUHKJGLHZEM-VIPPSAFOSA-L | |
disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate | |
44386827 | |
5 g | |
Allura Red AC |
C18H14N2Na2O8S2 | |
FD and C Red No.40 | |
[Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O | |
496.42 | |
496.42 | |
Crystal/Powder |
Safety and Handling
RTECSNumber : QK2260000
TSCA : Yes