missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Acetoacetate 95.0+%, TCI America™
$70.84 - $572.27
Chemical Identifiers
| CAS | 1118-84-9 |
|---|---|
| Molecular Formula | C7H10O3 |
| Molecular Weight (g/mol) | 142.154 |
| MDL Number | MFCD00009811 |
| InChI Key | AXLMPTNTPOWPLT-UHFFFAOYSA-N |
| Synonym | allyl acetoacetate, allyl 3-oxobutanoate, allyl acetylacetate, acetoacetic acid, allyl ester, butanoic acid, 3-oxo-, 2-propenyl ester, prop-2-en-1-yl 3-oxobutanoate, acetoacetic acid allyl ester, 2-propenyl 3-oxobutanoate, ac-allyl, unii-8hx066j62p |
| PubChem CID | 70701 |
| IUPAC Name | prop-2-enyl 3-oxobutanoate |
| SMILES | CC(=O)CC(=O)OCC=C |
Chemical Identifiers
| 1118-84-9 | |
| 142.154 | |
| AXLMPTNTPOWPLT-UHFFFAOYSA-N | |
| 70701 | |
| CC(=O)CC(=O)OCC=C |
| C7H10O3 | |
| MFCD00009811 | |
| allyl acetoacetate, allyl 3-oxobutanoate, allyl acetylacetate, acetoacetic acid, allyl ester, butanoic acid, 3-oxo-, 2-propenyl ester, prop-2-en-1-yl 3-oxobutanoate, acetoacetic acid allyl ester, 2-propenyl 3-oxobutanoate, ac-allyl, unii-8hx066j62p | |
| prop-2-enyl 3-oxobutanoate |
Specifications
| 1118-84-9 | |
| Yellow | |
| C7H10O3 | |
| 25 g | |
| allyl acetoacetate, allyl 3-oxobutanoate, allyl acetylacetate, acetoacetic acid, allyl ester, butanoic acid, 3-oxo-, 2-propenyl ester, prop-2-en-1-yl 3-oxobutanoate, acetoacetic acid allyl ester, 2-propenyl 3-oxobutanoate, ac-allyl, unii-8hx066j62p | |
| CC(=O)CC(=O)OCC=C | |
| 142.154 | |
| 142.15 | |
| Liquid |
| -85°C | |
| 195°C | |
| MFCD00009811 | |
| 2810 | |
| AXLMPTNTPOWPLT-UHFFFAOYSA-N | |
| prop-2-enyl 3-oxobutanoate | |
| 70701 | |
| ≥95.0% (GC) | |
| Allyl Acetoacetate |
Safety and Handling
EINECSNumber : (2)-3495
TSCA : No