missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Supplier:  TCI America A174025G

Catalog No. A174025G


Explore available promotions

Description

Description

Specifications

Specifications

Allyl Acrylate (stabilized with MEHQ)
Colorless
C6H8O2
25 g
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
C=CCOC(=O)C=C
112.128
112.13
Liquid
999-55-3
122°C
MFCD00014949
2929
QTECDUFMBMSHKR-UHFFFAOYSA-N
prop-2-enyl prop-2-enoate
13835
≥98.0% (GC)

Chemical Identifiers

999-55-3
112.128
QTECDUFMBMSHKR-UHFFFAOYSA-N
13835
C=CCOC(=O)C=C
C6H8O2
MFCD00014949
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
prop-2-enyl prop-2-enoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1001

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions