missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allyl Benzoate 98.0+%, TCI America™
Supplier: TCI America B2041250G
Specifications
Allyl Benzoate | |
Yellow | |
C10H10O2 | |
250 g | |
LYJHVEDILOKZCG-UHFFFAOYSA-N | |
prop-2-enyl benzoate | |
11406 | |
≥98.0% (GC) |
583-04-0 | |
242°C | |
MFCD00026104 | |
Benzoic Acid Allyl Ester | |
C=CCOC(=O)C1=CC=CC=C1 | |
162.188 | |
162.19 | |
Liquid |
Chemical Identifiers
583-04-0 | |
162.188 | |
LYJHVEDILOKZCG-UHFFFAOYSA-N | |
11406 | |
C=CCOC(=O)C1=CC=CC=C1 |
C10H10O2 | |
MFCD00026104 | |
Benzoic Acid Allyl Ester | |
prop-2-enyl benzoate |
Safety and Handling
EINECSNumber : (3)-1357
TSCA : No