Learn More
Allyl chloroacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1153106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allyl chloroacetate | |
1.16 | |
61°C (141°F) | |
1.445 | |
5 g | |
1747000 | |
VMBJJCDVORDOCF-UHFFFAOYSA-N | |
prop-2-enyl 2-chloroacetate | |
76206 | |
98% |
2916-14-5 | |
162°C to 164°C | |
C5H7ClO2 | |
MFCD00058936 | |
UN2810 | |
allyl chloroacetate, allyl 2-chloroacetate, chloroacetic acid allyl ester, acetic acid, chloro-, 2-propenyl ester, prop-2-en-1-yl 2-chloroacetate, acetic acid, chloro-, allyl ester, allyl monochloroacetate, acmc-1cffa, chloroacetic acid,2-propenyl ester | |
C=CCOC(=O)CCl | |
134.559 | |
134.56 |
Chemical Identifiers
2916-14-5 | |
134.559 | |
VMBJJCDVORDOCF-UHFFFAOYSA-N | |
76206 | |
C=CCOC(=O)CCl |
C5H7ClO2 | |
MFCD00058936 | |
allyl chloroacetate, allyl 2-chloroacetate, chloroacetic acid allyl ester, acetic acid, chloro-, 2-propenyl ester, prop-2-en-1-yl 2-chloroacetate, acetic acid, chloro-, allyl ester, allyl monochloroacetate, acmc-1cffa, chloroacetic acid,2-propenyl ester | |
prop-2-enyl 2-chloroacetate |
Safety and Handling
GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
H227-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 220-839-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only