missing translation for 'onlineSavingsMsg'
Learn More

Allyl iodide, 97%, stabilized

CAS: 556-56-9 | C3H5I | 167.98 g/mol

$202.22 - $636.68

Chemical Identifiers

CAS 556-56-9
Molecular Formula C3H5I
Molecular Weight (g/mol) 167.98
MDL Number MFCD00001094
InChI Key HFEHLDPGIKPNKL-UHFFFAOYSA-N
Synonym allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
PubChem CID 11166
IUPAC Name 3-iodoprop-1-ene
SMILES ICC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218540250
SDS View Documents
Thermo Scientific Chemicals
218540250
25 mL Glass bottle
Each for $202.22
Only null left
 
AC218541000
SDS View Documents
Thermo Scientific Chemicals
218541000
100 mL Glass bottle
Each for $636.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

556-56-9
167.98
HFEHLDPGIKPNKL-UHFFFAOYSA-N
11166
ICC=C
C3H5I
MFCD00001094
allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
3-iodoprop-1-ene

Specifications

556-56-9
Brown-Red or Orange-Red to Yellow
102.0°C to 103.0°C
Authentic
Glass bottle
1.5530 to 1.5550
MFCD00001094
01,202
15,285
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
ICC=C
167.98
11166
97%
Allyl iodide
-99.0°C
1.8300g/mL
18°C
96% min. (GC)
C3H5I
H2C=CHCH2I
25 mL
1.83
allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
HFEHLDPGIKPNKL-UHFFFAOYSA-N
3-iodoprop-1-ene
Copper
167.98
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 209-130-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only