missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 102951000

 View more versions of this product

Catalog No. AC102951000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Allyl isothiocyanate
57-06-7
100.0
White
151.0°C to 153.0°C
Authentic
Glass bottle
1.5290 to 1.5310
MFCD00004822
04, 214
15, 286
Solubility in water: 2g/L water (20°C). Other solubilities: ethanol of 65 vol%- 1:20 v/v,diethylphthalate - in all proportions,paraffin oil 65 cp- in all proportions,1,2-propanediol - 1:5 v/v,soluble in ether,acetone,benzene,cs2
C=CCN=C=S
99.15
5971
99.15
Fine Crystalline Powder
94%
92.0
-80.0°C
1.0175g/mL
46°C
92% min. (GC)
C4H5NS
H2C=CHCH2NCS
100 g
1.0175
allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
ZOJBYZNEUISWFT-UHFFFAOYSA-N
3-isothiocyanatoprop-1-ene
0.01% alpha-tocopherol
CHEBI:73224
94%

Chemical Identifiers

57-06-7
ZOJBYZNEUISWFT-UHFFFAOYSA-N
5971
3-isothiocyanatoprop-1-ene
MFCD00004822
allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
CHEBI:73224
C=CCN=C=S
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very t

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remov

GHS Signal Word: Danger

EINECSNumber : 200-309-2

RTECSNumber : NX8225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only