Learn More
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
$106.34 - $106.34
Chemical Identifiers
CAS | 1730-25-2 |
---|---|
Molecular Formula | C3H5BrMg |
Molecular Weight (g/mol) | 145.28 |
MDL Number | MFCD00000044 |
InChI Key | FEMBXICCJNZMMC-UHFFFAOYSA-M |
Synonym | allylmagnesium bromide, grignard reagent, allyl magnesium bromide, sjpdabpprujp@, sjpdabpprujp@, allylmagnesiumbromid, allylmagnesiumbromid, allyl magnesiumbromide, allyl magnesiumbromide, 2-propenylmagnesium bromide, 2-propenylmagnesium bromide |
PubChem CID | 74413 |
IUPAC Name | magnesium;prop-1-ene;bromide |
SMILES | Br[Mg]CC=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC209531000
|
Thermo Scientific Chemicals
209531000 |
100 mL | AcroSeal™ Glass bottle |
Each for $106.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1730-25-2 | |
145.28 | |
FEMBXICCJNZMMC-UHFFFAOYSA-M | |
74413 | |
Br[Mg]CC=C |
C3H5BrMg | |
MFCD00000044 | |
allylmagnesium bromide, grignard reagent, allyl magnesium bromide, sjpdabpprujp@, sjpdabpprujp@, allylmagnesiumbromid, allylmagnesiumbromid, allyl magnesiumbromide, allyl magnesiumbromide, 2-propenylmagnesium bromide, 2-propenylmagnesium bromide | |
magnesium;prop-1-ene;bromide |
Specifications
1730-25-2,1730-25-2 | |
18.78,84.64 | |
0.8510g/mL | |
AcroSeal™ Glass bottle | |
H2C=CHCH2MgBr | |
100 mL | |
0.851 | |
FEMBXICCJNZMMC-UHFFFAOYSA-M | |
magnesium;prop-1-ene;bromide | |
74413 | |
Solution |
15.37,81.22 | |
Gray or Green-Yellow | |
−40°C | |
C3H5BrMg | |
MFCD00000044 | |
3399 | |
allylmagnesium bromide, grignard reagent, allyl magnesium bromide, sjpdabpprujp@, sjpdabpprujp@, allylmagnesiumbromid, allylmagnesiumbromid, allyl magnesiumbromide, allyl magnesiumbromide, 2-propenylmagnesium bromide, 2-propenylmagnesium bromide | |
Br[Mg]CC=C | |
145.28 | |
145.28 | |
Allylmagnesium bromide, 1M solution in diethyl ether |
Safety and Handling
GHS H Statement
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if swallowed.
Causes severe skin bur
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 217-046-4
RUO – Research Use Only