Learn More
Allyltriphenylphosphonium bromide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1449030
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allyltriphenylphosphonium bromide | |
222°C to 225°C | |
(CH2=CHCH2)(C6H5)3PBr | |
250 g | |
Hygroscopic | |
FWYKRJUVEOBFGH-UHFFFAOYSA-M | |
triphenyl(prop-2-enyl)phosphanium;bromide | |
197740 | |
99% |
1560-54-9 | |
C21H20BrP | |
MFCD00011808 | |
3579053 | |
allyltriphenylphosphonium bromide, triphenyl allylphosphonium bromide, phosphonium, triphenyl-2-propenyl-, bromide, allyl triphenylphosphonium bromide, phosphonium, allyltriphenyl-, bromide, allyltriphenylphosphonium bromide tal, triphenyl prop-2-enyl phosphanium bromide, 2-propenyltriphenylphosphonium bromide, triphenyl prop-2-en-1-yl phosphanium bromide, wln: 1u2pr&r&r &e | |
[Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
383.27 | |
383.27 |
Chemical Identifiers
1560-54-9 | |
383.27 | |
FWYKRJUVEOBFGH-UHFFFAOYSA-M | |
197740 | |
[Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C21H20BrP | |
MFCD00011808 | |
allyltriphenylphosphonium bromide, triphenyl allylphosphonium bromide, phosphonium, triphenyl-2-propenyl-, bromide, allyl triphenylphosphonium bromide, phosphonium, allyltriphenyl-, bromide, allyltriphenylphosphonium bromide tal, triphenyl prop-2-enyl phosphanium bromide, 2-propenyltriphenylphosphonium bromide, triphenyl prop-2-en-1-yl phosphanium bromide, wln: 1u2pr&r&r &e | |
triphenyl(prop-2-enyl)phosphanium;bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-332-6
RTECSNumber : TA1843000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only