Learn More
United Scientific™ Alnico U-Shaped Magnet
Powerful magnet for a variety of classroom applications.
Supplier:  United Scientific™ AUM025
Catalog No. S15777
Only null left
Explore available promotions
Description
Description
  • This powerful U-shaped Alnico magnet is painted red and supplied with a keeper.
  • 1 in. Red U-Shaped Alnico Magnet
  • North Pole marked with a dot
  • Sold in EA
Specifications
Specifications
Alnico
Magnetic Demonstrations
5+
Magnet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm