Learn More
alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals
$195.86 - $527.13
Chemical Identifiers
CAS | 623-24-5 |
---|---|
Molecular Formula | C8H8Br2 |
Molecular Weight (g/mol) | 263.95 |
MDL Number | MFCD00000182 |
InChI Key | RBZMSGOBSOCYHR-UHFFFAOYSA-N |
Synonym | 1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775 |
PubChem CID | 69335 |
IUPAC Name | 1,4-bis(bromomethyl)benzene |
SMILES | C1=CC(=CC=C1CBr)CBr |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC112940250
|
Thermo Scientific Chemicals
112940250 |
25 g | Glass bottle |
Each for $195.86
|
|
||||
AC112941000
|
Thermo Scientific Chemicals
112941000 |
100 g | Glass bottle |
Each for $527.13
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
623-24-5 | |
263.95 | |
RBZMSGOBSOCYHR-UHFFFAOYSA-N | |
69335 | |
C1=CC(=CC=C1CBr)CBr |
C8H8Br2 | |
MFCD00000182 | |
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775 | |
1,4-bis(bromomethyl)benzene |
Specifications
623-24-5 | |
100.0 | |
White to Beige | |
245.0°C | |
96% min. (GC) | |
C8H8Br2 | |
MFCD00000182 | |
05, 385 | |
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775 | |
RBZMSGOBSOCYHR-UHFFFAOYSA-N | |
1,4-bis(bromomethyl)benzene | |
69335 | |
97% | |
α,α′-Dibromo-p-xylene, 97% |
96.0 | |
142.0°C to 147.0°C | |
2.0120g/mL | |
Authentic | |
Glass bottle | |
C6H4(CH2Br)2 | |
25 g | |
2.012 | |
Solubility in water: soluble,hydrolyses | |
C1=CC(=CC=C1CBr)CBr | |
263.95 | |
263.95 | |
Crystalline Powder and Coarse Crystals |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 210-781-1
TSCA : TSCA
RUO – Research Use Only