missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dimethyl-gamma-butyrolactone 97.0+%, TCI America™

$719.52 - $2468.02

Chemical Identifiers

CAS 3709-08-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD03844788
InChI Key UPVAIJPDWVTFKT-UHFFFAOYSA-N
Synonym Dihydro-3,3-dimethyl-2(3H)-furanone
PubChem CID 94746
IUPAC Name 3,3-dimethyloxolan-2-one
SMILES CC1(CCOC1=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D27335G
View Documents
TCI America
D27335G
5 g
Each for $719.52
Only null left
 
D273325G
View Documents
TCI America
D273325G
25 g
Each for $2,468.02
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3709-08-8
114.14
UPVAIJPDWVTFKT-UHFFFAOYSA-N
94746
CC1(CCOC1=O)C
C6H10O2
MFCD03844788
Dihydro-3,3-dimethyl-2(3H)-furanone
3,3-dimethyloxolan-2-one

Specifications

3709-08-8
Yellow
C6H10O2
5 g
UPVAIJPDWVTFKT-UHFFFAOYSA-N
3,3-dimethyloxolan-2-one
94746
≥97.0% (GC)
alpha,alpha-Dimethyl-gamma-butyrolactone
6°C
194°C
MFCD03844788
Dihydro-3,3-dimethyl-2(3H)-furanone
CC1(CCOC1=O)C
114.14
114.14
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'