missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Benzaldoxime 90.0+%, TCI America™
$167.76 - $1,321.62
Chemical Identifiers
| CAS | 622-31-1 |
|---|---|
| Molecular Formula | C7H7NO |
| Molecular Weight (g/mol) | 121.139 |
| MDL Number | MFCD00002119 |
| InChI Key | VTWKXBJHBHYJBI-SOFGYWHQSA-N |
| Synonym | benzaldehyde oxime, benzaldoxime, benzaldehyde, oxime, e-benzaldehyde oxime, alpha-benzaldoxime, benzaldehydeoxime, syn-benzaldoxime, benzaldehyde, oxime, e, e-n-phenylmethylidene hydroxylamine, z-benzaldoxime |
| PubChem CID | 5324611 |
| IUPAC Name | (NE)-N-benzylidenehydroxylamine |
| SMILES | C1=CC=C(C=C1)C=NO |
Chemical Identifiers
| 622-31-1 | |
| 121.139 | |
| VTWKXBJHBHYJBI-SOFGYWHQSA-N | |
| 5324611 | |
| C1=CC=C(C=C1)C=NO |
| C7H7NO | |
| MFCD00002119 | |
| benzaldehyde oxime, benzaldoxime, benzaldehyde, oxime, e-benzaldehyde oxime, alpha-benzaldoxime, benzaldehydeoxime, syn-benzaldoxime, benzaldehyde, oxime, e, e-n-phenylmethylidene hydroxylamine, z-benzaldoxime | |
| (NE)-N-benzylidenehydroxylamine |
Specifications
| 622-31-1 | |
| 123°C | |
| MFCD00002119 | |
| benzaldehyde oxime, benzaldoxime, benzaldehyde, oxime, e-benzaldehyde oxime, alpha-benzaldoxime, benzaldehydeoxime, syn-benzaldoxime, benzaldehyde, oxime, e, e-n-phenylmethylidene hydroxylamine, z-benzaldoxime | |
| C1=CC=C(C=C1)C=NO | |
| 121.139 | |
| 121.14 | |
| Crystalline Lumps |
| White-Yellow | |
| C7H7NO | |
| 25 g | |
| VTWKXBJHBHYJBI-SOFGYWHQSA-N | |
| (NE)-N-benzylidenehydroxylamine | |
| 5324611 | |
| ≥90.0% (GC) | |
| alpha-Benzaldoxime |
Safety and Handling
TSCA : Yes