Learn More
Thermo Scientific Chemicals alpha-Chloralose, 98+%, beta anomer ca 20%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1149214
Description
It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canadensis) and blue geese (Chen caerulescens) were caught with alpha-chloralose, an oral hypnotic administered on bait. Wild turkeys (Meleagris gallopavo) were captured with alpha-chloralose, an oral anesthetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canaden
Specifications
alpha-Chloralose | |
beta anomer ≈ 15% | |
25 g | |
MFCD00005542 | |
14,2072 | |
Slightly soluble in water. | |
OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl | |
309.52 | |
309.53 |
180°C to 184°C | |
15879-93-3 | |
C8H11Cl3O6 | |
85418 | |
a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol | |
OJYGBLRPYBAHRT-OPKHMCHVSA-N | |
1-[(2R,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol | |
7057995 | |
≥98% |
Chemical Identifiers
15879-93-3 | |
309.52 | |
OJYGBLRPYBAHRT-OPKHMCHVSA-N | |
7057995 |
C8H11Cl3O6 | |
MFCD00005542 | |
a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol | |
OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl |
Safety and Handling
GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
H301-H332-H336
EINECSNumber : 240-016-7
RTECSNumber : FM9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only