missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
alpha-D-Lactose monohydrate, Thermo Scientific Chemicals
$51.01 - $471.00
Chemical Identifiers
CAS | 5989-81-1 |
---|---|
Molecular Formula | C12H24O12 |
Molecular Weight (g/mol) | 360.312 |
MDL Number | MFCD00150747 |
InChI Key | WSVLPVUVIUVCRA-KPKNDVKVSA-N |
Synonym | alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x |
PubChem CID | 104938 |
IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate |
SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1107430
|
Thermo Scientific Chemicals
A1107430 |
250 g |
Each for $51.01
|
|
|||||
AAA110740B
|
Thermo Scientific Chemicals
A110740B |
1000 g |
Each for $120.85
|
|
|||||
AAA110740I
|
Thermo Scientific Chemicals
A110740I |
5000 g |
Each for $471.00
|
|
|||||
Description
alpha-D-Lactose monohydrate is used as a carrier and stabiliser of aromas, pharmaceutical products, Food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Chemical Identifiers
5989-81-1 | |
360.312 | |
WSVLPVUVIUVCRA-KPKNDVKVSA-N | |
104938 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O |
C12H24O12 | |
MFCD00150747 | |
alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate |
Specifications
5989-81-1 | |
1.525 | |
C12H24O12 | |
250 g | |
14,5343 | |
Soluble in water. | |
+52° (c=4.5 in water, 22h) | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate | |
104938 | |
alpha-D-Lactose monohydrate |
207°C to 211°C (decomposition) | |
Odorless | |
MFCD00150747 | |
3768231 | |
alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x | |
WSVLPVUVIUVCRA-KPKNDVKVSA-N | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O | |
360.312 | |
360.32 (342.30 Anhydrous) |
Safety and Handling
EINECSNumber : 200-559-2
RTECSNumber : OD9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only