Learn More
alpha-Ionone, 90+%, Thermo Scientific Chemicals
$87.75 - $305.19
Chemical Identifiers
CAS | 127-41-3 |
---|---|
Molecular Formula | C13H20O |
Molecular Weight (g/mol) | 192.30 |
MDL Number | MFCD00001565 |
InChI Key | UZFLPKAIBPNNCA-BQYQJAHWNA-N |
Synonym | alpha-ionone, iraldeine, e-alpha-ionone, irisone, alpha-ionon, trans-alpha-ionone, alpha-e-ionone, alpha-ionone natural, beta-lonone, ionone, alpha |
PubChem CID | 5282108 |
ChEBI | CHEBI:32319 |
IUPAC Name | (E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one |
SMILES | CC(=O)\C=C\C1C(C)=CCCC1(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1709822
|
Thermo Scientific Chemicals
A1709822 |
100 g |
Each for $87.75
|
|
|||||
AAA1709836
|
Thermo Scientific Chemicals
A1709836 |
500 g |
Each for $305.19
|
|
|||||
Description
alpha-Ionone, is used in many floral accords. Gives a violet flower nuance to compositions, but is also used in rose, woody, amber, balsamic, and powdery fragrances. It is also used in perfumery, flavoring, and vitamin A (retinol) production for cosmetics and toiletries. beta isomer shows a less floral odor than the alpha. It has good substantivity and is used in all areas of perfumery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsalpha-Ionone, is used in many floral accords. Gives a violet flower nuance to compositions, but is also used in rose, woody, amber, balsamic, and powdery fragrances. It is also used in perfumery, flavoring, and vitamin A (retinol) production for cosmetics and toiletries. beta isomer shows a less floral odor than the alpha. It has good substantivity and is used in all areas of perfumery.
Solubility
Soluble in ethanol fixed oils, propylene glycol. Slightly soluble in alcohol, ether, mineral oil.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
127-41-3 | |
192.30 | |
UZFLPKAIBPNNCA-BQYQJAHWNA-N | |
5282108 | |
(E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one |
C13H20O | |
MFCD00001565 | |
alpha-ionone, iraldeine, e-alpha-ionone, irisone, alpha-ionon, trans-alpha-ionone, alpha-e-ionone, alpha-ionone natural, beta-lonone, ionone, alpha | |
CHEBI:32319 | |
CC(=O)\C=C\C1C(C)=CCCC1(C)C |
Specifications
127-41-3 | |
237°C | |
C13H20O | |
MFCD00001565 | |
2046084 | |
alpha-ionone, iraldeine, e-alpha-ionone, irisone, alpha-ionon, trans-alpha-ionone, alpha-e-ionone, alpha-ionone natural, beta-lonone, ionone, alpha | |
UZFLPKAIBPNNCA-BQYQJAHWNA-N | |
(E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one | |
5282108 | |
192.3 | |
alpha-Ionone |
0.931 | |
117°C (243°F) | |
1.498 | |
100 g | |
14,5056 | |
Soluble in ethanol fixed oils,propylene glycol. Slightly soluble in alcohol,ether,mineral oil. | |
CC(=O)\C=C\C1C(C)=CCCC1(C)C | |
192.30 | |
CHEBI:32319 | |
≥90% |
Safety and Handling
EINECSNumber : 204-841-6
RTECSNumber : EN0525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only