Learn More
alpha-Methylstyrene, 99%, stab. with 10-20 ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
$65.74 - $217.32
Chemical Identifiers
CAS | 98-83-9 |
---|---|
Molecular Formula | C9H10 |
Molecular Weight (g/mol) | 118.18 |
MDL Number | MFCD00008859 |
InChI Key | XYLMUPLGERFSHI-UHFFFAOYSA-N |
Synonym | alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene |
PubChem CID | 7407 |
ChEBI | CHEBI:35060 |
IUPAC Name | prop-1-en-2-ylbenzene |
SMILES | CC(=C)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL03609AE
|
Thermo Scientific Chemicals
L03609AE |
100 mL |
Each for $65.74
|
|
||||||
AAL03609AP
|
Thermo Scientific Chemicals
L03609AP |
500 mL |
Each for $79.26
|
|
||||||
AAL036090F
|
Thermo Scientific Chemicals
L036090F |
2500 mL |
Each for $217.32
|
|
||||||
Description
α-Methylstyrene (AMS) is used in the synthesis of (acrylonitrile-styrene-acrylic copolymer)/(α-methylstyrene-acrylonitrile copolymer). It is used in the manufacture of plasticizers, resins and polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsα-Methylstyrene (AMS) is used in the synthesis of (acrylonitrile-styrene-acrylic copolymer)/(α-methylstyrene-acrylonitrile copolymer). It is used in the manufacture of plasticizers, resins and polymers.
Solubility
Soluble in water.( 0,1 g/L) at 25°C.
Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents, Peroxides, Organometallic compounds, Metallic salts.
Chemical Identifiers
98-83-9 | |
118.18 | |
XYLMUPLGERFSHI-UHFFFAOYSA-N | |
7407 | |
prop-1-en-2-ylbenzene |
C9H10 | |
MFCD00008859 | |
alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene | |
CHEBI:35060 | |
CC(=C)C1=CC=CC=C1 |
Specifications
98-83-9 | |
0.909 | |
45°C (113°F) | |
1.538 | |
100 mL | |
969405 | |
Soluble in water.( 0,1g/L) at 25°C. | |
CC(=C)C1=CC=CC=C1 | |
118.18 | |
CHEBI:35060 | |
99% |
-23°C | |
166°C to 169°C | |
C9H10 | |
MFCD00008859 | |
UN2303 | |
alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene | |
XYLMUPLGERFSHI-UHFFFAOYSA-N | |
prop-1-en-2-ylbenzene | |
7407 | |
118.18 | |
alpha-Methylstyrene, stabilized with 10 to 20ppm 4-tert-butylcatechol |
Safety and Handling
GHS H Statement
H226-H319-H335
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P333+P313-P363-P370+P378q-P501c
H226-H304-H317-H319-H335-H336-H351-H361
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOPROPENYLBENZENE
EINECSNumber : 202-705-0
RTECSNumber : WL5075300
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only