Alpha Scientific™ DIFF-SAFE™ Blood Dispenser for Blood Smears
Cuts the time of smear-making in half, reducing technician fatigue
$78.86 - $13540.71
Specifications
Product Type | Blood Dispenser |
---|---|
Includes | 21G x 0.5 in. Blunt needle |
For Use With (Application) | Blood smears |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
22-029115
|
Alpha Scientific™
DS101PK |
100 Pack |
Pack of 100 Each for $78.86
|
|
|||||
22-026396
|
Alpha Scientific™
DS101 |
Case of 10000 |
Pack of 1000 Each for $704.00
|
|
|||||
22-026397
|
Alpha Scientific™
DS125 |
25000 Case |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Cuts the time of smear-making in half, reducing technician fatigue
- Compared to the typical procedure involving 15 or more steps when sticks or capillaries are used, the procedure using the DIFF-SAFE dispenser cuts the number of steps by two thirds
- Blunt cannula size is 21G × 0.5 in.
Specifications
Blood Dispenser | |
21G x 0.5 in. Blunt needle | |
Blood smears |