missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-Terpineol, 96%, Thermo Scientific Chemicals

A naturally-occuring monoterpene alcohol

$60.26 - $204.53

Chemical Identifiers

CAS 98-55-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00001557
InChI Key WUOACPNHFRMFPN-UHFFFAOYSA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1628514
View Documents
Thermo Scientific Chemicals
01628514
25 g
Each for $60.26
 
AA1628522
View Documents
Thermo Scientific Chemicals
01628522
100 g
Each for $105.00
 
AA1628536
View Documents
Thermo Scientific Chemicals
01628536
500 g
Each for $204.53
 
Description

Description

A naturally-occuring monoterpene alcohol.Alpha-terpineol is used as an antioxidant, antiseptic, antihypernociception and anti-inflammatory. It is also used as a solvent. It is an important ingredient of pine oil disinfectants. Further, it is used as a fragrance in perfumes, fat denaturant for soap production and synthetic flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-55-5
154.253
WUOACPNHFRMFPN-UHFFFAOYSA-N
17100
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
C10H18O
MFCD00001557
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
CHEBI:22469
CC1=CCC(CC1)C(C)(C)O

Specifications

98-55-5
Colorless to Yellow
210°C to 219°C
C10H18O
MFCD00001557
14,9171
Miscible with water,benzene,propylene glycol,acetone,alcohol and ether.
CC1=CCC(CC1)C(C)(C)O
154.253
CHEBI:22469
96%
alpha-Terpineol
31°C to 32°C
0.930 g/mL
90°C (194°F)
1.483
25 g
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
WUOACPNHFRMFPN-UHFFFAOYSA-N
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
17100
154.24
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 202-680-6

RTECSNumber : WZ6700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only