missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals (±)-alpha-Tocopherol, 95%, synthetic

Supplier:  Thermo Scientific Chemicals 428120250

 View more versions of this product

Catalog No. AC428120250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(±)-α-Tocopherol
10191-41-0
100.0
0.9500g/mL
94% min. (HPLC)
C29H50O2
25 g
15, 9653
Solubility in water: insoluble. Other solubilities: freely soluble in oils, fats, acetone, alcohol,, chloroform, diethyl ether, other fat solvents.
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
430.72
4200 mPa.s (20°C)
430.71
Synthetic
94.0
2.5°C to 3.5°C
393.0°C
Glass bottle
MFCD00072051
0.95
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
GVJHHUAWPYXKBD-IEOSBIPESA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
14985
CHEBI:18145
95%

Chemical Identifiers

10191-41-0
430.72
GVJHHUAWPYXKBD-IEOSBIPESA-N
14985
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
C29H50O2
MFCD00072051
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
CHEBI:18145
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 233-466-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only