AlphaProTech Critical Cover™ UltraGrip™ Shoe Covers
Anti-skid shoe covers are extremely clean, durable, and strong to withstand abrasive floors. BarrierTech™ material provides particle holdout and protection from fluids and light chemical splash.
Supplier:  AlphaProTech SHM1316BH
 View more versions of this product
Catalog No. 19-130-1481
Explore available promotions
Description
Description
  • Ultimate in slip and fall protection on damp and dry floors
  • High performance, anti-skid properties
  • Full coverage up through ankle area
  • BarrierTech™ upper material provides protection from fluids and light chemical splash
  • Two colors, four sizes
  • Different colors around the opening provide convenient size identification
Specifications
Specifications
Large
Blue
Non-sterile
Orange
Orange
12.75 in.
5 in.
Dry Particulates, Fluid, Light Chemical Splash
Polyethylene, Polypropylene
Single-use
Anti-skid
Serged
12.75 x 4.5 x 5 in. (32.38 x 11.43 x 12.7cm)
32.38 cm
12.7 cm
Controlled Environments Shoe Covers