AlphaProTech Critical Cover™ UltraGrip™ Shoe Covers
Anti-skid shoe covers are extremely clean, durable, and strong to withstand abrasive floors. BarrierTech™ material provides particle holdout and protection from fluids and light chemical splash.
Supplier:  AlphaProTech SHM1313BH
 View more versions of this product
Catalog No. 19-130-1483
Explore available promotions
Description
Description
  • Ultimate in slip and fall protection on damp and dry floors
  • High performance, anti-skid properties
  • Full coverage up through ankle area
  • BarrierTech™ upper material provides protection from fluids and light chemical splash
  • Two colors, four sizes
  • Different colors around the opening provide convenient size identification
Specifications
Specifications
X-Large, X-Large
Blue, Blue
Non-sterile, Non-sterile
Black, Black
Black, Black
15 in., 15 in.
5.75 in., 5.75 in.
Dry Particulates, Fluid, Light Chemical Splash, Dry Particulates, Fluid, Light Chemical Splash
Polyethylene, Polypropylene
Single-use, Single-use
Anti-skid, Anti-skid
Serged, Serged
15 x 4.75 x 5.75 in. (38.1 x 12 x 14.6cm), 15 x 4.75 x 5.75 in. (38.1 x 12 x 14.6cm)
38.1 cm, 38.1 cm
14.6 cm, 14.6 cm
Controlled Environments Shoe Covers, Controlled Environments Shoe Covers