AlphaProTech Critical Cover™ UltraGrip™ Shoe Covers
Anti-skid shoe covers are extremely clean, durable, and strong to withstand abrasive floors. BarrierTech™ material provides particle holdout and protection from fluids and light chemical splash.
Supplier:  AlphaProTech SHE1312BH
 View more versions of this product
Catalog No. 29-635-3345
Explore available promotions
Description
Description
  • Ultimate in slip and fall protection on damp and dry floors
  • High performance, anti-skid properties
  • Full coverage up through ankle area
  • BarrierTech™ upper material provides protection from fluids and light chemical splash
  • Two colors, four sizes
  • Different colors around the opening provide convenient size identification
Specifications
Specifications
Universal, Universal
White, White
Non-sterile, Non-sterile
White, White
White, White
14 in., 14 in.
5 in., 5 in.
Dry Particulates, Dry Particulates, Fluid, Light Chemical Splash, Fluid, Light Chemical Splash
Polyethylene, Polypropylene, Polyethylene, Polypropylene
Single-use, Single-use
Anti-skid, Anti-skid
Serged, Serged
14 x 4.5 x 5 in. (35.56 x 11.43 x 12.7cm), 14 x 4.5 x 5 in. (35.56 x 11.43 x 12.7cm)
35.56 cm, 35.56 cm
12.7 cm, 12.7 cm
Controlled Environments Shoe Covers, Controlled Environments Shoe Covers