AlphaProTech Critical Cover™ GenPro™ Hood
Designed to protect against dirt, grime, and particulates in nonhazardous environments.
Supplier: AlphaProTech SD91342B
Description
- Lightweight, breathable material
- Comfortable, nonbinding latex-free elastic bands around face and neck for secure fit
- Provide secure complete coverage and comfort without pinching or binding
- Open face style provides complete coverage without obstructing vision or range of motion
Specifications
Combination Head and Beard Cover | |
Polypropylene | |
Elastic | |
Serged | |
21C CFR Part 820 | |
Bouffant and Beard Cover |
X-Large | |
White | |
Yes | |
Non-sterile | |
500/Cs. |