AlphaProTech Critical Cover™ AlphaGuard™ Frocks
Frocks are constructed with low particle count multilayer SMS material to offer high levels of breathability without compromising filtration efficiency.
Supplier:  AlphaProTech FK321227
 View more versions of this product
Catalog No. 19-130-1465G
Explore available promotions
Description
Description
  • Low particle count
  • Fluid repellent
  • Clean, durable and soft
  • Static dissipative
  • Raglan sleeves
  • Snap collar and close
  • Serged seams
Specifications
Specifications
Controlled Environments Frock
SMS
Non-sterile
Elastic
Mandarin
74.9 cm
104.1 cm
4X-Large
White
Snaps
Serged
29.5 in.
Raglan Sleeve