missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell AlphaTec™ 04-002 Medium Weight PVC Gloves 12 in. With Fleece Liner

Highly durable 12 in. long PVC gloves are ideal for food handling, chemical handling, maintenance and cleaning.

Supplier:  Ansell 04002100

 View more versions of this product

Catalog No. 19-500-992



Explore available promotions

Description

Description

  • FDA and EU food compliances
  • Oil resistant coating
  • Special rough grip for best dry, wet and oil grips
  • Handshape ergonomic design
Specifications

Specifications

ANSI 4
EN 420:2003 + A1:2009
Blue
Yes
10
No
JKL
Gauntlet
Fleece
04002100
12 in.
21 CFR 170-199
No
ANSI A2
PVC, Nitrile
EN 388:3121B; EN 388:3121
Type B
Bacteria, Fungi only
Rough
Lined
AlphaTec™ 04-414
30 cm
Videos
SDS
Documents

Documents

Promotions

Promotions