missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell AlphaTec™ 58-530B Liquid-Proof, Chemical Resistant Glove

Chemical protective and oil-resistant waterproof gauntlet gloves offer a confident grip and a comfortable inner lining. Ideal for outdoor applications.

Supplier:  Ansell 58530B100

 View more versions of this product

Catalog No. 17-999-824



Explore available promotions

Description

Description

  • Combination of liquid proof chemical resistance and grip, with flexibility and dexterity
  • Reliable chemical protection for added user safety.
  • ANSELL GRIP™ technology ensures that the coating polymer does not penetrate the liner during manufacture
  • Coating is consistent and sound, significantly reducing the likelihood of chemical leakage onto the skin
  • Recommended for outdoor applications
Specifications

Specifications

ANSI 5
EN 420:2003 + A1:2009, Category lll, EN 1149
Gray, Teal
Nitrile
EN 388:3122A, EN 388:4121
Type A
Bacteria, Fungi only
ANSELL GRIP™ Technology
Lined
AlphaTec™ 58-530B
29 to 31 cm
21 CFR 170-199
No
Yes
10
No
JKT
Gauntlet
Acrylic
58530B100
11 to 12 in.
Videos
SDS
Documents

Documents

Promotions

Promotions