missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell™ AlphaTec™ Medium Duty Splash Protection Vinyl Apron
Ansell™ AlphaTec™ Medium Duty Splash Protection Vinyl Apron keeps workers keep clean and protected from harmful contact wherever oil, grease and chemicals are present
$2.09 - $2.09
Specifications
Type | Apron |
---|---|
Material | PVC |
Color | Blue |
Closure Type | Adjustable Ties |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
19-010-251
|
Ansell™
ECB45 |
33 x 44 in. |
Each for $2.09
Case of 72 Each for $150.48
|
|
|||||
Description
- PVC (Vinyl) aprons traditionally offer excellent protection where splashes from acids and caustic bases are a concern
- Die-cut PVC construction features superior flexibility yet excellent resistance to abrasion and puncture
- Integral ties provide an economical solution for adjustable fit
- Applications:
- Packing of fillets and ground meat
- Canning
- Bottling
- Storing and transportation
- Equiptment maintenance and repair
- Plant disinfection and sanitization
- Transferring liquids
Specifications
Apron | |
Blue | |
Non-sterile | |
No |
PVC | |
Adjustable Ties | |
Die cut PVC | |
6 mil |