Learn More
Ansell™ Alphatec™ MICROCHEM™ 68-1500 PLUSFR Coveralls with Hood
Designed for protection from particulates and light, non-flammable liquid splashes. Ansell™ Alphatec™ MICROCHEM™ 68-1500 PLUSFR Coveralls with Hood are made of highly breathable, flame retardant and anti-static SMMS. They are perfect for work in utilities, industrial cleaning, general maintenance, and the petrochemical industry.
$291.88 - $367.46
Specifications
Material | FR SMMS |
---|---|
Color | Navy, Red |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic, Finger Loops |
Alert:
This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-999-996
|
Ansell™
NR17S9211102 |
Small |
Case of 25 Each for $291.88
|
|
|||||
18-000-000
|
Ansell™
NR17S9211106 |
2X-Large |
Case of 25 Each for $306.47
|
|
|||||
18-000-001
|
Ansell™
NR17S9211107 |
3X-Large |
Case of 25 Each for $309.32
|
|
|||||
17-999-997
|
Ansell™
NR17S9211103 |
Medium |
Case of 25 Each for $291.88
|
|
|||||
17-999-999
|
Ansell™
NR17S9211105 |
X-Large |
Case of 25 Each for $291.88
|
|
|||||
17-999-998
|
Ansell™
NR17S9211104 |
Large |
Case of 25 Each for $291.88
|
|
|||||
18-000-002
|
Ansell™
NR17S9211108 |
4X-Large |
Case of 25 Each for $334.12
|
|
|||||
18-000-003
|
Ansell™
NR17S9211109 |
5X-Large |
Case of 25 Each for $367.46
|
|
|||||
Description
- Protection: Flame retardant and anti-static SMMS nonwoven provides a good barrier to particulates and low hazard liquid splashes
- Comfort: Air and water vapor permeable (“breathable”) to help reduce the risk of heat stress
- Optimized body fit: Ensures full freedom of movement when worn over heat and flame protective clothing (EN ISO 14116 Index 2 or above)
- Anti-static: Tested according to EN1149-5
- Design: Elastic waist, wrists and ankles, 2-way front zipper with taped storm flap, finger loops, red stitching for ease of identification, serged seams, 3 piece respirator fit hood
- Applications: Petrochemical industry, industrial cleaning, utilities, general maintenance
- Sizes: Available in Small through 5X-Large
- Certifications: EN 1073-2, EN ISO 14116 / INDEX 1/0/0, EN 1149-5 / AATCC 76, EN ISO 13982-1 Type 5, ANSI/ISEA 101, ISO 17491-4 Type 6
- Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1073-2, EN ISO 14116 / INDEX 1/0/0, EN 1149-5 / AATCC 76, EN ISO 13982-1 Type 5, ANSI/ISEA 101, ISO 17491-4 Type 6
Specifications
FR SMMS | |
Elastic | |
Elastic, Finger Loops | |
3-panel, Respirator Fit | |
Serged | |
No |
Navy, Red | |
Elastic | |
Hooded | |
2-way Front Zipper with Storm Flap | |
Particulate and light liquid splash protection |