Ansell Alphatec™ 1800 Standard Bound Model 111 Coveralls with Hood
Coveralls with hood provide entry level, low hazard liquid and particulate protection for a wide range of general industrial applications. Previously known as MICROCHEM™ by AlphaTec 1800.
Supplier:  Ansell WH18B9211109
 View more versions of this product
Catalog No. 17-999-987
Explore available promotions
Alert:
This product contains natural rubber latex which may cause allergic reactions.
Description
Description
  • Provides low hazard liquid spray, splash and particle protection
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Anti-static, tested according to EN1149-5 and AATCC76 (8EMSL test method)
  • Elastic waist, wrists, and ankles
  • 2-way front zipper with storm flap
  • Bound seams, attached hood
  • Available in Small through 5X-Large sizes
  • Certifications EN 1149-5, AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 13
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
3-panel, Respirator Fit
Bound
Non-toxic Particulates, Liquid, Anti-static
EN 1149-5, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
WH18-B-92-111
General Maintenance, Paint Spraying, Powder Handling, Food
5X-Large
Microporous Film Laminate
Limited-use
Elastic
Hooded
Front Zipper with Storm Flap
Non-sterile
AATCC 76, ANSI/ISEA 101
No
None