missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell™ Alphatec™ MICROCHEM™ 68-2000 White Coveralls with Collar
Provide breathable protection and comfort with exceptional liquid and particulate protection. Ansell™ Alphatec™ MICROCHEM™ 68-2000 White Coveralls with Collar are ideal for a wide range of industrial applications.
Supplier: Ansell™ WH20B9210305
Description
- Protection: Excellent liquid penetration resistance & barrier to fine particulates (0.01μm) including ASTMF1761 and EN14126:2003 resistance to penetration of blood, body fluids and blood-borne pathogens
- Comfort: Moisture vapor permeable ("breathable") to help reduce the risk of heat stress
- Low Linting: Reduces the risk of fiber contamination in some critical areas
- Anti-static: Tested according to EN1149-5 & AATCC76 (8EMSL test method)
- Design: Elastic wrists and ankles, 2-way front zipper with storm flap, bound seams, finger loops
- Applications: Pharmaceutical industries, agriculture, cleanrooms, paint spraying, crime scene investigations, veterinary services, food
- Sizes: Available in Small through 5X-Large
- Certifications: EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
- Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
Specifications
X-Large | |
White | |
Elastic | |
Collared | |
Bound | |
Free of natural rubber latex |
Microporous Film Laminate | |
Elastic | |
Elastic | |
2-way Front Zipper with Storm Flap | |
Particulate and light liquid splash protection |