missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood
Provide strong, durable protection suitable for workers in extremely hazardous areas, including HAZMAT reponse teams. Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood reach new levels in chemical protection and have been engineered to protect.
Supplier: Ansell™ OR50T9211102
Alert:
This product contains natural rubber latex which may cause allergic reactions.
Description
- Protection: Barrier to numerous organic and inorganic chemicals and biological hazards; >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Comfort: Multi-layer material which is lightweight, yet strong and durable
- Highly visible: Bright orange color for improved worker safety
- Design: Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, ultrasonically welded and taped seams, 2 piece hood
- Applications: Industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, pharmaceuticals, mining, agriculture, HAZMAT emergency reponse (Level B), first response and fire service
- Sizes: Available in Small through 5X-Large
- Certifications: EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
- Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
Specifications
Small | |
Orange | |
Elastic | |
Hooded | |
Double Zipper | |
Liquid and spray tight chemical and biological protection | |
No |
Multi-layer Barrier Laminate Fabric | |
Elastic | |
Elastic, Knit | |
2-panel | |
Welded, Taped | |
Elasticated outer cuff with knitted inner cuff |