missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood

Provide strong, durable protection suitable for workers in extremely hazardous areas, including HAZMAT reponse teams. Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood reach new levels in chemical protection and have been engineered to protect.

Supplier:  Ansell™ OR50T9211107

 View more versions of this product

Catalog No. 17-999-945


Alert:

This product contains natural rubber latex which may cause allergic reactions.


Description

Description

  • Protection:  Barrier to numerous organic and inorganic chemicals and biological hazards; >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Comfort:  Multi-layer material which is lightweight, yet strong and durable
  • Highly visible:  Bright orange color for improved worker safety
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, ultrasonically welded and taped seams, 2 piece hood
  • Applications:  Industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, pharmaceuticals, mining, agriculture, HAZMAT emergency reponse (Level B), first response and fire service
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Specifications

Specifications

3X-Large
Orange
Elastic
Hooded
Double Zipper
Liquid and spray tight chemical and biological protection
No
Multi-layer Barrier Laminate Fabric
Elastic
Elastic, Knit
2-panel
Welded, Taped
Elasticated outer cuff with knitted inner cuff
SDS
Documents

Documents

Promotions

Promotions