missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ PVC Coat Apron

Ansell™ AlphaTec™ PVC Coat Apron keeps workers clean and protected wherever oil, grease and chemicals are present

$250.30 - $250.30

Spécifications

Material PVC
Color Blue
Disposable Reusable
Closure Type Ties
Sterility Non-sterile
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. Size Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Size Prix Quantité  
19-010-242
Afficher les documents
Ansell™
CAS8
Small
caisse de 12 chaque for $250.30
Il en reste null
 
19-010-243
Afficher les documents
Ansell™
CAM8
Medium
caisse de 12 chaque for $250.30
Il en reste null
 
19-010-244
Afficher les documents
Ansell™
CAL8
Large
caisse de 12 chaque for $250.30
Il en reste null
 
19-010-245
Afficher les documents
Ansell™
CAXL8
X-Large
caisse de 12 chaque for $250.30
Il en reste null
 
Description

Description

  • No exposed elastic or metal parts to cause static or corrosion
  • Protection when splashes from acids and caustic bases are a concern
  • Virgin vinyl construction contains no reprocessed materials
  • Flexible
  • Resistant to punctures, abrasion, chemicals, fats, oils and greases
  • Sleeves come with elasticized cuffs for added protection
  • PVC (vinyl) grommets are permanently sealed to the aprons for durability and allow for size adjustment without tear-out
  • Six sizes with adjustable ties ensure optimal fit for a wide range of users
Spécifications

Spécifications

PVC
Reusable
Non-sterile
No
Packing of filets and ground meat; Canning; Bottling; Storing and transportation; Equipment maintenance and repair; Plant disinfection and sanitization; Transferring liquids
Blue
Ties
Elastic
8 mil
Dielectrically Sealed
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'