missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alumina, Activity: II-III, MP EcoChrom™, MP Biomedicals™

Use MP EcoChrom™ Alumina, Activity II-III as an adsorbent, or in chromatography and thin layer chromatography (TLC).

Supplier:  MP Biomedicals Inc 0204691

 View more versions of this product

Catalog No. ICN04691


Explore available promotions

Description

Description

  • MP EcoChrom™ Alumina, Activity II-III is an economical adsorbents of medium activity.
  • It is often used where activated charcol can not be used due to its organic nature, or, where the cation exchange properties of basic alumina are desired.
  • It is suitable for the development of separation methods and for technical separations where a custom-made adsorbents for special separation is not available.
  • An oxide of aluminum, it occurs in nature as various minerals such as bauxite, corundum, etc.
  • It is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories.
Specifications

Specifications

Alumina
Pink to White
Free-flowing Powder
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Activity: II-III, Activity acc to Brockmann: 17/mm
1344-28-1
5 kg
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
3.94 g/cm3 (at 20°C)

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.