missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alumina B, Activity: I, MP EcoChrom™, MP Biomedicals™
MP EcoChrom™ Alumina B, Activity I are good all-round adsorbents for laboratory use as well as for industrial use. They can be adjusted to lower activity by the addition of polar media, preferably water, but also higher alcohols, (e.g. glycol or glycerol can be used).
Supplier: MP Biomedicals Inc 0202069
Description
- MP EcoChrom™ Alumina B, Activity I is an oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc.
- It is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories.
Specifications
Alumina B | |
Pink to White | |
Powder | |
Al2O3 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 |
Activity: I, Activity acc. To Brockmann: 24 l/mm | |
1344-28-1 | |
500 g | |
MFCD00003424 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
3.94 g/cm3 (at 20°C) |
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Safety and Handling
EINECSNumber : 215-691-6
Recommended Storage : Store at room temperature.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.