missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alumina B, Activity: Super I, Basic pH, MP EcoChrom™, MP Biomedicals™

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc. This Alumina is produced at the highest activity available in the market.

$364.96 - $8977.57

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN04571
View Documents
MP Biomedicals Inc
0204571
1 kg
Each for $364.96
 
ICN04574
View Documents
MP Biomedicals Inc
0204574
5 kg
Each for $1,263.85
 
ICN04577
View Documents
MP Biomedicals Inc
0204577
50 kg
Each for $8,977.57
 
Description

Description

  • MP EcoChrom™ Alumina - Super I is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.
  • MP Alumina B - Super I show absorption capacity approximately twice as high as an Alumin B, Activity I.
  • The initial activity is kept in a very narrow range and their deactivation behaviour is extremely constant.
  • The product is available as a neutral version, MP Alumina N - Super I, and an acidic version, MP Alumina A - Super I.
  • MP EcoChro Alumina B - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides, environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.
Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Pink to White
Free-flowing Powder
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Alumina B, Activity: Super I
1344-28-1
1 kg
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
9989226
3.94 g/cm3 (at 20°C)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

Recommended Storage : Store at room temperature.

SDS
missing translation for 'documents'

missing translation for 'documents'

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.