missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alumina N, Activity: I, MP EcoChrom™, MP Biomedicals™

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.

Supplier:  MP Biomedicals Inc 0202087

 View more versions of this product

Catalog No. ICN02087


Explore available promotions

Description

Description

  • MP EcoChrom™ Alumina is widely used as an adsorbent, desiccant, abrasive, electrical insulator and as a catalyst for various organic reactions.
  • It is also used for isolation of alkaloids, glycosides, active substances from plants, vitamins, hormones and antibiotics, dehydration of organic solvents, purification of enzymes, removal of terpenes, peroxides, pyrogens from injectable solutions and infusions.
Specifications

Specifications

Alumina B
Pink to White
Free-flowing Powder
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Activity: I
1344-28-1
1 kg
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
9989226
3.94 g/cm3 (at 20°C)

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.