missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alumina R, MP EcoChrom™, MP Biomedicals™

MP EcoChrom™ Alumin R is an acid alumina, developed for use in isotope chemistry.

$240.99 - $240.99

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN06034
View Documents
MP Biomedicals Inc
0206034
500 g
Each for $240.99
 
Description

Description

  • MP EcoChrom™ Alumina R catalyses a variety of reactions that are useful industrially.
  • It is widely used to remove water from gas streams.
  • Also used for its hardness and strength.
  • It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
  • Many types of sandpaper use aluminium oxide crystals.
Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminium(3+) trioxidandiide

Specifications

ca. 2072 °C
1344-28-1
Free-flowing Powder
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
9989226
At 20 °C (not applicable)
Alumina R
Pink to White
4.3 ± 0.1
500 g
MFCD00003424
Insoluble in water.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
3.94 g/cm3 (at 20°C)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

RTECSNumber : BD1200000

Recommended Storage : Store at room temperature.

SDS
missing translation for 'documents'

missing translation for 'documents'

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.