missing translation for 'onlineSavingsMsg'
Learn More

Aluminum bromide, 98+%, extra pure, anhydrous

CAS: 7727-15-3 | AlBr3 | 266.69 g/mol

$106.39 - $282.08

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC198410250
SDS View Documents
Thermo Scientific Chemicals
198410250
25 g Glass bottle
Each for $106.39
Only null left
 
AC198411000
SDS View Documents
Thermo Scientific Chemicals
198411000
100 g Glass bottle
Each for $282.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

7727-15-3
266.69
PQLAYKMGZDUDLQ-UHFFFAOYSA-K
[Al+3].[Br-].[Br-].[Br-]
AlBr3
MFCD00003421
aluminium(3+) tribromide

Specifications

265.0°C
7727-15-3
Crystalline Solid
98% min. (Complexometry)
AlBr3
01,22; 02,19; 03,7; 04,10; 05,10; 06,17; 09,11
Solubility in water: reacts. Other solubilities: soluble in alcohol, carbon disulfide, benzene,, toluene, ether, nitrobenzene and xylene
[Al+3].[Br-].[Br-].[Br-]
266.69
≥98%
Glass bottle
97.0°C
Gray or White
25 g
AlBr3
MFCD00003421
15,328
PQLAYKMGZDUDLQ-UHFFFAOYSA-K
aluminium(3+) tribromide
266.7
Extra Pure
Aluminum bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-779-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only