missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, 99%, extra pure, anhydrous, granules

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol

$122.22 - $182.03

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC195785000
SDS View Documents
Thermo Scientific Chemicals
195785000
500 g Glass bottle
Each for $122.22
Only null left
 
AC195780010
SDS View Documents
Thermo Scientific Chemicals
195780010
1 kg Glass bottle
Each for $182.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7446-70-0
133.33
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum(3+) trichloride
AlCl3
MFCD00003422
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

194.0°C
Yellow
500 g
AlCl3
MFCD00003422
15, 333
Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform
[Al+3].[Cl-].[Cl-].[Cl-]
133.33
99%
Glass bottle
2.4400g/mL
7446-70-0
Granules
98.5% min. (Argentometry)
AlCl3
01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum(3+) trichloride
133.34
Extra Pure
2.44
Aluminum chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 231-208-1

RTECSNumber : BD0525000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only