missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum potassium sulfate dodecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

$38.29 - $705.10

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlKO8S2·12H2O
Molecular Weight (g/mol) 474.39
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423260050
SDS View Documents
Thermo Scientific Chemicals
423260050
5 g Glass Bottle
Each for $38.29
 
AC423265000
SDS View Documents
Thermo Scientific Chemicals
423265000
500 g Plastic bottle
Each for $207.90
 
AC423260025
SDS View Documents
Thermo Scientific Chemicals
423260025
2.5 kg Plastic bottle
Each for $705.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7784-24-9
474.39
GNHOJBNSNUXZQA-UHFFFAOYSA-J
62667
aluminum;potassium;disulfate;dodecahydrate
AlKO8S2·12H2O
MFCD00149143
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
CHEBI:86465
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

92°C
White
5 g
AlKO8S2·12H2O
MFCD00149143
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
GNHOJBNSNUXZQA-UHFFFAOYSA-J
aluminum;potassium;disulfate;dodecahydrate
62667
474.39
ACS Reagent
Aluminum potassium sulfate dodecahydrate
7784-24-9
Crystals
98.0 to 102% (Complexometry)
AlK(SO4)2·12H2O
15, 354
Solubility in water: 140g/L (20°C). Other solubilities: freely soluble in glycerol,insoluble in alcohol
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]
474.39
CHEBI:86465
≥98%
Glass Bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

RTECSNumber : WS5690000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only