Learn More
Aluminum acetate, basic hydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1162022
Description
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.
Solubility
Insoluble in water
Notes
Incompatible with strong oxidizing agents.
Specifications
Aluminum Acetate, Basic Hydrate | |
142-03-0 | |
C4H7AlO5 | |
14,344 | |
Insoluble in water | |
CC(=O)O[Al](O)OC(C)=O | |
162.08 |
Basic hydrate | |
100 g | |
MFCD00008688 | |
aluminum acetate, basic hydrate, c4h7alo5.h2o, aluminum diacetate hydrate | |
HQQUTGFAWJNQIP-UHFFFAOYSA-K | |
diacetyloxyaluminum;hydrate | |
18502856 |
Chemical Identifiers
142-03-0 | |
162.08 | |
HQQUTGFAWJNQIP-UHFFFAOYSA-K | |
18502856 |
C4H7AlO5 | |
MFCD00008688 | |
aluminum acetate, basic hydrate, c4h7alo5.h2o, aluminum diacetate hydrate | |
CC(=O)O[Al](O)OC(C)=O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-518-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only